Assay Details
Target Gene Details
Entrez Gene ID: | 72077 |
Gene Name: | glucosaminyl (N-acetyl) transferase 3, mucin type |
Gene Aliases: |
2010013H22Rik, 2210021I22Rik, 2210401J11Rik |
Location: |
Chr.9:70031496-70039887 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcnt3 | NM_028087.2 | 2 | 283 | NP_082363.2 |
| EF202835.1 | 2 | 283 |
More Information
Set Membership: |
|

Genomic Map