Assay Details
Target Gene Details
Entrez Gene ID: | 70993 |
Gene Name: | protease, serine 54 |
Gene Aliases: |
4931432M23Rik, Klkbl4 |
Location: |
Chr.8:95559067-95576330 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss54 | NM_027640.1 | 5 | 1059 | NP_081916.1 |
| AK016501.1 | 7 | 1376 | ||
| BC144859.1 | 6 | 1186 | ||
| BC145787.1 | 6 | 1186 |
More Information
Set Membership: |
|

Genomic Map