Assay Details
Target Gene Details
Entrez Gene ID: | 17961 |
Gene Name: | N-acetyltransferase 2 (arylamine N-acetyltransferase) |
Gene Aliases: |
AV377607 |
Location: |
Chr.8:67494875-67502644 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat2 | NM_001168577.1 | 2 | 922 | NP_001162049.1 |
| NM_010874.3 | 2 | 1020 | NP_035004.1 | |
| AJ251710.1 | 2 | 1020 | CAC12846.1 | |
| AK007583.1 | 2 | 931 | ||
| BC012972.1 | 2 | 940 | AAH12972.1 | |
| U35886.1 | 1 | 1104 | AAA78943.1 | |
| U35887.1 | 1 | 1104 | AAA78944.1 |
More Information
Set Membership: |
|

Genomic Map