Assay Details
Target Gene Details
Entrez Gene ID: | 235599 |
Gene Name: | RIKEN cDNA 6430571L13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:107340640-107349683 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 6430571L13Rik | NM_175486.3 | 6 | 1677 | NP_780695.1 |
| AK032358.1 | 6 | 1678 | BAC27833.1 | |
| BC115775.1 | 7 | 1560 | ||
| BC116708.1 | 7 | 1560 |

Genomic Map