Assay Details
Target Gene Details
Entrez Gene ID: | 69771 |
Gene Name: | RIKEN cDNA 1810019D21 gene |
Gene Aliases: |
- |
Location: |
Chr.8:106135400-106138723 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810019D21Rik | NR_040344.1 | |||
| NR_040345.1 | ||||
| NR_040346.1 |
Target Gene Details
Entrez Gene ID: | 77411 |
Gene Name: | epithelial splicing regulatory protein 2 |
Gene Aliases: |
9530027K23Rik, Rbm35b |
Location: |
Chr.8:106130403-106137528 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrp2 | NM_176838.2 | 1 | 137 | NP_789808.1 |
| AK088755.1 | ||||
| AK168488.1 | BAE40375.1 | |||
| BC031444.1 | 1 | 137 | AAH31444.1 |
More Information
Set Membership: |
|

Genomic Map