Assay Details
Target Gene Details
Entrez Gene ID: | 68865 |
Gene Name: | ARV1 homolog (yeast) |
Gene Aliases: |
1110067L22Rik, AI461928, AW121084 |
Location: |
Chr.8:124722138-124740648 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arv1 |
Target Gene Details
Entrez Gene ID: | 69627 |
Gene Name: | family with sequence similarity 89, member A |
Gene Aliases: |
2310031A18Rik |
Location: |
Chr.8:124740257-124751809 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam89a | NM_001081120.1 | 2 | 978 | NP_001074589.1 |
| AK009550.1 | 1 | 552 | ||
| BC023460.1 | 2 | 819 | AAH23460.1 |
More Information
Set Membership: |
|

Genomic Map