Assay Details
Target Gene Details
Entrez Gene ID: | 234542 |
Gene Name: | retbindin |
Gene Aliases: |
A330096I21Rik, BC022744, Rtbnd |
Location: |
Chr.8:84946263-84956606 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rtbdn | NM_144929.2 | 6 | 1706 | NP_659178.1 |
AK039726.1 | 4 | 2213 | ||
AK080715.1 | 6 | 1024 | ||
AY527275.1 | 6 | 1706 | AAS20324.1 | |
AY527276.1 | 7 | 1161 | AAS20325.1 | |
BC022744.1 | 5 | 1278 | AAH22744.1 | |
BC023051.1 | 5 | 1234 | AAH23051.1 | |
BC027079.1 | 5 | 1234 | AAH27079.1 |
