Assay Details
Target Gene Details
Entrez Gene ID: | 30947 |
Gene Name: | adenosine deaminase, tRNA-specific 1 |
Gene Aliases: |
MMADAT1, mADAT1 |
Location: |
Chr.8:111966897-111992307 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adat1 | NM_013925.4 | 10 | 2891 | NP_038953.1 |
| AK082399.1 | 10 | 2892 | BAC38487.1 |
More Information
Set Membership: |
|

Genomic Map