Assay Details
Target Gene Details
Entrez Gene ID: | 71764 |
Gene Name: | C2 calcium-dependent domain containing 2-like |
Gene Aliases: |
1300006O23Rik, Tmem24 |
Location: |
Chr.9:44309237-44320324 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2cd2l | NM_027909.2 | 14 | 2648 | NP_082185.2 |
| AK004920.1 | 14 | 2653 | BAB23671.1 | |
| AK122237.1 | 9 | 4223 | BAC65519.1 | |
| AK153804.1 | 14 | 2620 | BAE32190.1 | |
| BC035271.1 | 11 | 2654 | AAH35271.1 | |
| BC049875.1 | 10 | 1575 | AAH49875.1 | |
| BC056612.1 | 14 | 2219 | AAH56612.1 | |
| BC060156.1 | 14 | 2512 | AAH60156.1 |

Genomic Map