Assay Details
Target Gene Details
Entrez Gene ID: | 18768 |
Gene Name: | protein kinase inhibitor beta, cAMP dependent, testis specific |
Gene Aliases: |
Pkibeta, Prkacn2 |
Location: |
Chr.10:57631981-57741112 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkib | NM_001039050.1 | NP_001034139.1 | ||
| NM_001039051.2 | NP_001034140.1 | |||
| NM_001039052.1 | NP_001034141.1 | |||
| NM_001039053.2 | NP_001034142.2 | |||
| NM_001190402.1 | NP_001177331.1 | |||
| NM_008863.4 | NP_032889.3 | |||
| AK005281.1 | ||||
| AK083875.1 | BAC39046.1 | |||
| AK156950.1 | BAE33910.1 | |||
| AK157521.1 | BAE34112.1 | |||
| AK163312.1 | BAE37294.1 | |||
| AK171359.1 | 4 | 1801 | ||
| BC061162.1 | AAH61162.1 | |||
| BC087894.1 | AAH87894.1 | |||
| BE648461.1 | ||||
| L02241.1 | AAB59678.1 |
More Information
Set Membership: |
|

Genomic Map