Assay Details
Target Gene Details
Entrez Gene ID: | 18768 |
Gene Name: | protein kinase inhibitor beta, cAMP dependent, testis specific |
Gene Aliases: |
Pkibeta, Prkacn2 |
Location: |
Chr.10:57631981-57741112 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkib | NM_001039050.1 | 5 | 2370 | NP_001034139.1 |
| NM_001039051.2 | 5 | 2176 | NP_001034140.1 | |
| NM_001039052.1 | 5 | 2036 | NP_001034141.1 | |
| NM_001039053.2 | 6 | 2072 | NP_001034142.2 | |
| NM_001190402.1 | 5 | 2274 | NP_001177331.1 | |
| NM_008863.4 | 6 | 2284 | NP_032889.3 | |
| AK083875.1 | 6 | 2185 | BAC39046.1 | |
| AK156950.1 | 5 | 2063 | BAE33910.1 |
More Information
Set Membership: |
|

Genomic Map