Assay Details
Target Gene Details
Entrez Gene ID: | 69836 |
Gene Name: | phospholipase A2, group XIIB |
Gene Aliases: |
2010002E04Rik, Fksg71, Pla2g13, hlb218 |
Location: |
Chr.10:59403420-59421976 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g12b | NM_023530.2 | 1 | 247 | NP_076019.2 |
| AF339738.1 | 1 | 269 | AAK01739.1 | |
| AK008035.1 | 1 | 273 | ||
| AY358033.1 | 1 | 276 | AAQ92919.1 | |
| BC021592.1 | 1 | 247 | AAH21592.1 |
More Information
Set Membership: |
|

Genomic Map