Assay Details
Target Gene Details
Entrez Gene ID: | 21376 |
Gene Name: | transforming growth factor beta regulated gene 1 |
Gene Aliases: |
AA408552, AA409675, Niam, Tb-5, mFLJ00213 |
Location: |
Chr.9:37649182-37657312 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbrg1 | NM_025289.3 | 9 | 1461 | NP_079565.2 |
| AK003270.1 | 9 | 1387 | ||
| AK010925.1 | 9 | 1411 | ||
| AK011268.1 | 9 | 1386 | ||
| AK131158.1 | 6 | 3671 | BAD21408.1 | |
| AK150549.1 | 9 | 1388 | BAE29651.1 | |
| AK151075.1 | 9 | 1339 | BAE30090.1 | |
| AK151209.1 | 9 | 1355 | BAE30204.1 | |
| AK151457.1 | 9 | 1373 | BAE30416.1 | |
| AK166547.1 | 9 | 1383 | BAE38845.1 | |
| AK205100.1 | ||||
| BC021331.1 | 9 | 1472 | AAH21331.1 | |
| BC040813.1 | 9 | 1374 | ||
| BC043022.1 | 9 | 1376 | ||
| BC065795.1 | 9 | 1374 | AAH65795.1 | |
| BQ174234.1 | 1 | 123 |
More Information
Set Membership: |
|

Genomic Map