Assay Details
Target Gene Details
Entrez Gene ID: | 69519 |
Gene Name: | RWD domain containing 2A |
Gene Aliases: |
1700030C20Rik, AI848608, Rwdd2 |
Location: |
Chr.9:86571988-86574899 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rwdd2a | NM_001145968.1 | 3 | 883 | NP_001139440.1 |
| NM_001145969.1 | 3 | 917 | NP_001139441.1 | |
| NM_027100.2 | 3 | 1012 | NP_081376.2 | |
| AI848608.1 | 1 | 323 | ||
| AK006533.1 | 3 | 1011 | BAB24638.1 | |
| BC115691.1 | 3 | 1012 | ||
| BC115692.1 | 3 | 883 |
More Information
Set Membership: |
|

Genomic Map