Assay Details
Target Gene Details
Entrez Gene ID: | 77531 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 1B |
Gene Aliases: |
AIDA-1b, C030032C09Rik, E530015N03Rik, ENSMUSG00000078431, Gm10937, Gm1555 |
Location: |
Chr.10:89873509-90973300 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anks1b | NM_001128086.2 | 13 | 2013 | NP_001121558.2 |
More Information
Set Membership: |
|

Genomic Map