Assay Details
Target Gene Details
Entrez Gene ID: | 14806 |
Gene Name: | glutamate receptor, ionotropic, kainate 2 (beta 2) |
Gene Aliases: |
AW124492, GluK2, Glur-6, Glur6, Glurbeta2 |
Location: |
Chr.10:49094842-49789250 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grik2 | NM_001111268.1 | 17 | 3581 | NP_001104738.1 |
| NM_010349.2 | 18 | 3668 | NP_034479.2 | |
| AK034147.1 | 4 | 3542 | ||
| AK047667.1 | 2 | 725 | ||
| CF539412.1 | 1 | 445 |
More Information
Set Membership: |
|

Genomic Map