Assay Details
Target Gene Details
Entrez Gene ID: | 67454 |
Gene Name: | IKBKB interacting protein |
Gene Aliases: |
1200009F10Rik, 1700023M03Rik, D18354, Ikip |
Location: |
Chr.10:91083039-91102613 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbip | NM_027078.2 | 3 | 657 | NP_081354.1 |
| AK006283.1 | 3 | 625 | BAB24502.1 | |
| BC034890.1 | 3 | 619 | AAH34890.1 |

Genomic Map