Assay Details
Target Gene Details
Entrez Gene ID: | 103161 |
Gene Name: | apolipoprotein F |
Gene Aliases: |
AI255964 |
Location: |
Chr.10:128267997-128270151 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apof | NM_133997.2 | 1 | 383 | NP_598758.1 |
More Information
Set Membership: |
|

Genomic Map