Assay Details
Target Gene Details
Entrez Gene ID: | 12905 |
Gene Name: | CASP2 and RIPK1 domain containing adaptor with death domain |
Gene Aliases: |
RAIDD |
Location: |
Chr.10:95174739-95324345 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cradd | NM_009950.2 | 2 | 167 | NP_034080.1 |
| AF017992.1 | 1 | 48 | AAC35882.1 | |
| AJ224738.1 | 2 | 166 | CAA12107.1 | |
| AJ224740.1 | 2 | 152 | CAA12109.1 | |
| AK052304.1 | 2 | 72 | ||
| AK131713.1 | 2 | 274 | BAE20775.1 | |
| AK136561.1 | 2 | 72 | BAE23045.1 | |
| AK144648.1 | 2 | 188 | BAE25988.1 | |
| AK163112.1 | 2 | 179 | BAE37198.1 | |
| BC005608.1 | 1 | 130 | AAH05608.1 |

Genomic Map