Assay Details
Target Gene Details
Entrez Gene ID: | 74498 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
5430411C10Rik, GOLPH5, GRASP65, P65 |
Location: |
Chr.9:119925673-119937558 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gorasp1 | NM_028976.2 | 9 | 1904 | NP_083252.1 |
| AK017293.1 | 2 | 932 | BAB30676.1 | |
| AK154986.1 | 9 | 1881 | BAE32975.1 | |
| AK170627.1 | 9 | 1868 | BAE41921.1 | |
| BC012251.1 | 9 | 1904 | AAH12251.1 |

Genomic Map