Assay Details
Target Gene Details
Entrez Gene ID: | 270156 |
Gene Name: | expressed sequence AU019823 |
Gene Aliases: |
Gm639 |
Location: |
Chr.9:50605240-50617589 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU019823 | NM_001134902.1 | NP_001128374.1 | ||
| NM_212449.2 | NP_997614.2 | |||
| CJ142322.1 |
Target Gene Details
Entrez Gene ID: | 72614 |
Gene Name: | PIH1 domain containing 2 |
Gene Aliases: |
2700059L22Rik |
Location: |
Chr.9:50617321-50625000 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pih1d2 | NM_028300.2 | 1 | 141 | NP_082576.1 |
| AK012458.1 | 1 | 117 | BAB28253.1 | |
| AK080744.1 | 1 | 130 | ||
| BC019481.1 | 1 | 79 | ||
| BC039645.1 | 1 | 131 | AAH39645.1 |

Genomic Map