Assay Details
Target Gene Details
Entrez Gene ID: | 17172 |
Gene Name: | achaete-scute complex homolog 1 (Drosophila) |
Gene Aliases: |
AI225900, ASH1, Mash1, bHLHa46 |
Location: |
Chr.10:87491041-87493660 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascl1 | NM_008553.4 | 1 | 219 | NP_032579.2 |
| AK141866.1 | 1 | 209 | ||
| AK143210.1 | 1 | 220 | BAE25307.1 |
More Information
Set Membership: |
|

Genomic Map