Assay Details
Target Gene Details
Entrez Gene ID: | 73106 |
Gene Name: | protease, serine 57 |
Gene Aliases: |
2900092M14Rik, GLGL782, Prssl1, UNQ782 |
Location: |
Chr.10:79781474-79795605 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss57 | NM_001042710.1 | 5 | 1034 | NP_001036175.1 |
| AK013841.1 | 2 | 488 | ||
| AK153740.1 | 5 | 1002 | ||
| AK158376.1 | 5 | 1035 |
More Information
Set Membership: |
|

Genomic Map