Assay Details
Target Gene Details
Entrez Gene ID: | 270156 |
Gene Name: | expressed sequence AU019823 |
Gene Aliases: |
Gm639 |
Location: |
Chr.9:50605240-50617589 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU019823 | NM_001134902.1 | NP_001128374.1 | ||
| NM_212449.2 | NP_997614.2 | |||
| BC067205.1 | AAH67205.1 |
Target Gene Details
Entrez Gene ID: | 30057 |
Gene Name: | translocase of inner mitochondrial membrane 8B |
Gene Aliases: |
Tim8b |
Location: |
Chr.9:50603901-50605320 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timm8b | NM_013897.2 | 2 | 399 | NP_038925.1 |
| AA204614.1 | 2 | 388 | ||
| AK003382.1 | 2 | 358 | BAB22753.1 | |
| AK012925.1 | 2 | 366 | BAB28552.1 | |
| AK018718.1 | 2 | 366 | BAB31364.1 | |
| AK019422.1 | 2 | 374 | BAB31713.1 | |
| AK169258.1 | 2 | 378 | BAE41021.1 | |
| BC029239.1 | 2 | 352 | AAH29239.1 | |
| BF148992.1 |

Genomic Map