Assay Details
Target Gene Details
Entrez Gene ID: | 67341 |
Gene Name: | achaete-scute complex homolog 4 (Drosophila) |
Gene Aliases: |
1700054F16Rik, bHLHa44 |
Location: |
Chr.10:85928491-85929647 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascl4 | NM_001163614.1 | 1 | 420 | NP_001157086.1 |
More Information
Set Membership: |
|

Genomic Map