Assay Details
Target Gene Details
Entrez Gene ID: | 66403 |
Gene Name: | anti-silencing function 1A histone chaperone |
Gene Aliases: |
2310079C17Rik, C79052 |
Location: |
Chr.10:53596961-53609225 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asf1a | NM_025541.3 | 4 | 1930 | NP_079817.1 |
| AK031513.1 | 4 | 1896 | ||
| AK088618.1 | 4 | 1873 | BAC40457.1 |
Target Gene Details
Entrez Gene ID: | 71567 |
Gene Name: | minichromosome maintenance complex component 9 |
Gene Aliases: |
9030408O17Rik, Gm235, Mcmdc1 |
Location: |
Chr.10:53536315-53630439 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm9 | NM_027830.2 | NP_082106.2 | ||
| AK086212.1 | 1 | 1908 | ||
| AK139112.1 |
More Information
Set Membership: |
|

Genomic Map