Assay Details
Target Gene Details
Entrez Gene ID: | 16467 |
Gene Name: | ataxia, cerebellar, Cayman type homolog (human) |
Gene Aliases: |
3322401A10Rik, BB077577, BNIP-H, CLAC, hes, ji |
Location: |
Chr.10:81204513-81230779 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atcay | NM_178662.3 | 13 | 3299 | NP_848777.1 |
| AK038990.1 | 13 | 3300 | BAC30195.1 | |
| BC048903.1 | 13 | 3283 | AAH48903.1 |
Target Gene Details
Entrez Gene ID: | 69678 |
Gene Name: | ataxia, cerebellar, Cayman type homolog (human), opposite strand |
Gene Aliases: |
2310050B05Rik |
Location: |
Chr.10:81194691-81208370 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atcayos | NR_015477.1 | |||
| AK084504.1 |
More Information
Set Membership: |
|

Genomic Map