Assay Details
Target Gene Details
Entrez Gene ID: | 28015 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide M |
Gene Aliases: |
AA407243, AA408799, AI303746, AW553238, D9Wsu138e, Grinl1a, mKIAA4185 |
Location: |
Chr.9:71478437-71485983 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2m | NM_001164793.1 | 4 | 1618 | NP_001158265.1 |
NM_178602.3 | 4 | 1694 | NP_848717.1 | |
AK002571.1 | 4 | 1619 | BAB22196.1 | |
AK003433.1 | 2 | 694 | ||
AK014316.1 | 4 | 1626 | BAB29267.1 | |
AK017773.1 | 4 | 1637 | ||
AK051868.1 | 4 | 1628 | BAC34793.1 | |
AK052037.1 | 4 | 1648 | BAC34836.1 | |
AK078750.1 | 4 | 1629 | ||
AK079119.1 | 4 | 1612 | BAC37548.1 | |
AK136806.1 | 4 | 1646 | ||
AK151221.1 | 4 | 1632 | ||
AK159924.1 | 4 | 1641 | BAE35486.1 | |
AK166738.1 | 4 | 1641 | BAE38981.1 | |
AK206099.1 | 1 | 112 | ||
AK219410.1 | ||||
BC018502.1 | 4 | 1620 | AAH18502.1 | |
BC062199.1 | 4 | 1614 | AAH62199.1 |
