Assay Details
Target Gene Details
Entrez Gene ID: | 380629 |
Gene Name: | headcase homolog (Drosophila) |
Gene Aliases: |
Gm869, HDC, HDCL |
Location: |
Chr.10:17899042-17948067 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Heca | NM_001033432.3 | 4 | 3033 | NP_001028604.1 |
| AK132340.1 | 4 | 3034 | BAE21115.1 |

Genomic Map