Assay Details
Target Gene Details
Entrez Gene ID: | 15214 |
Gene Name: | hairy/enhancer-of-split related with YRPW motif 2 |
Gene Aliases: |
CHF1, Herp1, Hrt2, bHLHb32, hesr2 |
Location: |
Chr.10:30832359-30842783 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hey2 | NM_013904.1 | 5 | 1852 | NP_038932.1 |
| AB093589.1 | 5 | 1720 | BAC55066.1 | |
| AF172287.1 | 5 | 1852 | AAF14546.1 | |
| AF173902.1 | 5 | 1844 | AAF20174.1 | |
| AF232240.1 | 5 | 1848 | AAF37298.1 | |
| AJ271867.1 | 5 | 1852 | CAB71346.1 | |
| AK031506.1 | 5 | 1699 | BAC27428.1 | |
| AK158000.1 | 5 | 1853 | BAE34310.1 |
More Information
Set Membership: |
|

Genomic Map