Assay Details
Target Gene Details
Entrez Gene ID: | 21412 |
Gene Name: | transcription factor 21 |
Gene Aliases: |
Pod-1, Pod1, bHLHa23, capsulin, epc, epicardin |
Location: |
Chr.10:22817263-22820128 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcf21 | NM_011545.1 | 1 | 233 | NP_035675.1 |
| AB009453.1 | 1 | 230 | BAA23883.1 | |
| AF029753.1 | 1 | 220 | AAB84256.1 | |
| AF035717.1 | 1 | 233 | AAC62513.1 | |
| AF036945.1 | 1 | 271 | AAC23537.1 | |
| AF047418.1 | 1 | 206 | AAC62531.1 | |
| AK011575.1 | 1 | 249 | BAB27709.1 | |
| AK135423.1 | 1 | 280 | BAE22527.1 | |
| AK157289.1 | 1 | 249 | BAE34032.1 | |
| AK161795.1 | 1 | 265 | ||
| BC053525.1 | 1 | 240 | AAH53525.1 |
More Information
Set Membership: |
|

Genomic Map