Assay Details
Target Gene Details
Entrez Gene ID: | 216198 |
Gene Name: | t-complex 11 (mouse) like 2 |
Gene Aliases: |
E430026E19Rik |
Location: |
Chr.10:84576947-84614355 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcp11l2 | NM_146008.2 | 2 | 322 | NP_666120.1 |
| AK040850.1 | 2 | 373 | ||
| AK146035.1 | 2 | 645 | BAE26848.1 | |
| BC029663.1 | 2 | 322 | AAH29663.1 |
More Information
Set Membership: |
|

Genomic Map