Assay Details
Target Gene Details
Entrez Gene ID: | 15364 |
Gene Name: | high mobility group AT-hook 2 |
Gene Aliases: |
9430083A20Rik, HMGI-C, Hmgic, pg, pygmy |
Location: |
Chr.10:120361275-120476935 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmga2 | NM_010441.2 | 1 | 175 | NP_034571.1 |
| CJ173998.1 | 1 | 175 |
More Information
Set Membership: |
|

Genomic Map