Assay Details
Target Gene Details
Entrez Gene ID: | 94222 |
Gene Name: | oligodendrocyte transcription factor 3 |
Gene Aliases: |
Bhlhb7, bHLHe20 |
Location: |
Chr.10:19356539-19358604 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olig3 | NM_053008.2 | 1 | 691 | NP_443734.2 |
| AB038698.1 | 1 | 718 | BAB18908.1 | |
| BC057564.1 | 1 | 732 | AAH57564.1 |
More Information
Set Membership: |
|

Genomic Map