Assay Details
Target Gene Details
Entrez Gene ID: | 19272 |
Gene Name: | protein tyrosine phosphatase, receptor type, K |
Gene Aliases: |
AI853699, PTPk, RPTPkappa |
Location: |
Chr.10:28074331-28597397 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprk | NM_008983.2 | 1 | 292 | NP_033009.1 |
| AK019870.1 | 1 | 191 | ||
| AK052275.1 | 1 | 245 | ||
| AK078614.1 | 2 | 209 | BAC37341.1 | |
| AK083006.1 | 1 | 256 | BAC38727.1 | |
| BC138376.1 | 1 | 258 |
More Information
Set Membership: |
|

Genomic Map