Assay Details
Target Gene Details
Entrez Gene ID: | 319880 |
Gene Name: | transmembrane and coiled coil domains 3 |
Gene Aliases: |
AW488095, C630016B22Rik, C88213, Tmcc1 |
Location: |
Chr.10:94311912-94590954 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmcc3 | NM_001168684.1 | 4 | 1927 | NP_001162155.1 |
| NM_172051.3 | 4 | 2004 | NP_742048.2 | |
| NM_177026.1 | 4 | 1946 | NP_796000.1 | |
| AK079611.1 | 5 | 1864 | BAC37701.1 | |
| AK083129.1 | 4 | 1907 | BAC38774.1 | |
| AK132545.1 | 4 | 1982 | BAE21229.1 | |
| BC026867.1 | 4 | 1927 | AAH26867.1 | |
| BC057202.1 | 4 | 1790 | AAH57202.1 | |
| BC095951.1 | 4 | 1938 | AAH95951.1 |

Genomic Map