Assay Details
Target Gene Details
Entrez Gene ID: | 71090 |
Gene Name: | Sumo1 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.10:51756174-51757194 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933411G06Rik | NR_045158.1 | 1 | 336 | |
| AK016770.1 | 1 | 337 | BAB30417.1 |

Genomic Map