Assay Details
Target Gene Details
Entrez Gene ID: | 16000 |
Gene Name: | insulin-like growth factor 1 |
Gene Aliases: |
C730016P09Rik, Igf-1, Igf-I |
Location: |
Chr.10:87858270-87937047 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf1 | NM_001111274.1 | 1 | 89 | NP_001104744.1 |
| NM_001111275.1 | NP_001104745.1 | |||
| NM_001111276.1 | 1 | 89 | NP_001104746.1 | |
| NM_010512.4 | NP_034642.2 | |||
| NM_184052.3 | 1 | 89 | NP_908941.1 | |
| AK038119.1 | 1 | 1171 | BAC29934.1 | |
| AK050118.1 | 1 | 2133 | ||
| AK052033.1 | 1 | 1935 | ||
| AK081019.1 | BAC38117.1 | |||
| AK155435.1 | BAE33264.1 | |||
| AK165471.1 | BAE38208.1 | |||
| AY878192.1 | AAX61179.1 | |||
| AY878193.1 | AAX61180.1 |
More Information
Set Membership: |
|

Genomic Map