Assay Details
Target Gene Details
Entrez Gene ID: | 237360 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 14 |
Gene Aliases: |
Adamts-14, TS14 |
Location: |
Chr.10:61197112-61273438 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts14 | NM_001081127.1 | 22 | 3961 | NP_001074596.1 |
| AK137573.1 | 7 | 1413 | BAE23412.1 | |
| AK145103.1 | ||||
| BC157953.1 | 22 | 3796 | ||
| BC158014.1 | 22 | 3794 |
More Information
Set Membership: |
|

Genomic Map