Assay Details
Target Gene Details
Entrez Gene ID: | 74007 |
Gene Name: | BTB (POZ) domain containing 11 |
Gene Aliases: |
6330404E16Rik |
Location: |
Chr.10:85386776-85660292 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btbd11 | NM_028709.2 | 1 | 276 | NP_082985.2 |
More Information
Set Membership: |
|

Genomic Map