Assay Details
Target Gene Details
Entrez Gene ID: | 74007 |
Gene Name: | BTB (POZ) domain containing 11 |
Gene Aliases: |
6330404E16Rik |
Location: |
Chr.10:85386776-85660292 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btbd11 | NM_001017525.1 | 15 | 2849 | NP_001017525.1 |
| NM_028709.2 | 17 | 4702 | NP_082985.2 | |
| AK018115.1 | 7 | 2163 | BAB31077.1 | |
| AK039038.1 | 14 | 2645 | BAC30216.1 | |
| AK044022.1 | 15 | 2740 | BAC31741.1 | |
| BC059036.1 | 5 | 1720 | AAH59036.1 | |
| BC072592.1 | 17 | 3327 | AAH72592.1 |
More Information
Set Membership: |
|

Genomic Map