Assay Details
Target Gene Details
Entrez Gene ID: | 52665 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 1 |
Gene Aliases: |
1700028A24Rik, AI314462, AI930038, D10Ertd667e, MMCD |
Location: |
Chr.10:29311507-29346661 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Echdc1 | NM_001110195.1 | 6 | 1012 | NP_001103665.1 |
| NM_025855.4 | 6 | 1114 | NP_080131.4 | |
| AK003965.1 | 1 | 361 | BAB23096.1 | |
| AK006444.1 | 6 | 991 | BAB24592.2 | |
| AK016260.1 | 6 | 1055 | ||
| AK028775.1 | 6 | 1011 | BAC26112.1 | |
| AK150932.1 | 6 | 996 | BAE29969.1 | |
| AK152285.1 | 6 | 996 | BAE31098.1 | |
| AK153454.1 | 6 | 996 | BAE32007.1 | |
| AK166589.1 | 6 | 1036 | BAE38876.1 | |
| AK166634.1 | 6 | 1036 | BAE38907.1 | |
| AK166660.1 | 6 | 1036 | BAE38924.1 | |
| BC066183.1 | 6 | 979 | AAH66183.1 |
Target Gene Details
Entrez Gene ID: | 68031 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
2610509H23Rik, Iduna |
Location: |
Chr.10:29344176-29362442 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf146 | NM_001110196.1 | 3 | 3792 | NP_001103666.1 |
| NM_001110197.1 | 4 | 3853 | NP_001103667.1 | |
| NM_001110198.1 | 4 | 3833 | NP_001103668.1 | |
| NM_001284279.1 | 3 | 3365 | NP_001271208.1 | |
| NM_026518.4 | 3 | 3772 | NP_080794.2 | |
| AK021080.1 | 1 | 838 | BAB32294.1 | |
| AK035665.1 | 1 | 1180 | BAC29145.1 |
More Information
Set Membership: |
|

Genomic Map