Assay Details
Target Gene Details
Entrez Gene ID: | 237504 |
Gene Name: | Ras association (RalGDS/AF-6) domain family (N-terminal) member 9 |
Gene Aliases: |
AW322107, Pamci |
Location: |
Chr.10:102512222-102549737 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf9 | NM_146240.4 | 2 | 950 | NP_666352.1 |
| AK033464.1 | 2 | 754 | BAC28302.1 | |
| AK041688.1 | 2 | 951 | BAC31033.1 | |
| AK086855.2 | 2 | 886 | ||
| BC028805.1 | 2 | 887 | AAH28805.1 |
More Information
Set Membership: |
|

Genomic Map