Assay Details
Target Gene Details
Entrez Gene ID: | 66663 |
Gene Name: | ubiquitin-like modifier activating enzyme 5 |
Gene Aliases: |
5730525G14Rik, AW240750, Ube1dc1 |
Location: |
Chr.9:104046588-104063176 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uba5 | NM_025692.3 | 12 | 1495 | NP_079968.2 |
| AK011151.1 | 1 | 55 | ||
| AK017787.1 | BAB30933.1 | |||
| AK077825.1 | 12 | 1476 | ||
| AK151823.1 | BAE30720.1 | |||
| AK152029.1 | 12 | 1490 | BAE30888.1 | |
| AK168907.1 | 12 | 1489 | BAE40721.1 | |
| AK197371.1 | ||||
| BC019764.1 | AAH19764.1 |
More Information
Set Membership: |
|

Genomic Map