Assay Details
Target Gene Details
Entrez Gene ID: | 11540 |
Gene Name: | adenosine A2a receptor |
Gene Aliases: |
A2AAR, A2aR, AA2AR |
Location: |
Chr.10:75316860-75334792 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adora2a | NM_009630.3 | 2 | 640 | NP_033760.2 |
| AK206618.1 | ||||
| BC110692.1 | 1 | 140 | AAI10693.1 | |
| CJ135665.1 | 2 | 574 | ||
| CX241487.1 | 2 | 445 | ||
| U05672.1 | 1 | 366 | AAA19000.1 | |
| Y13346.1 | 1 | 125 | CAA73788.1 |
More Information
Set Membership: |
|

Genomic Map