Assay Details
Target Gene Details
Entrez Gene ID: | 104248 |
Gene Name: | calcineurin binding protein 1 |
Gene Aliases: |
A330070M20Rik, Cain, Ppp3in |
Location: |
Chr.10:75646110-75764358 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cabin1 | NM_172549.3 | 28 | 5038 | NP_766137.2 |
| AK039606.1 | 2 | 741 | ||
| AK078480.1 | BAC37299.1 | |||
| BC057551.1 | AAH57551.1 | |||
| BC150825.1 | 27 | 4602 | ||
| CV884170.1 | 3 | 529 |
More Information
Set Membership: |
|

Genomic Map