Assay Details
Target Gene Details
Entrez Gene ID: | 67500 |
Gene Name: | cell division cycle and apoptosis regulator 1 |
Gene Aliases: |
2610511G16Rik, 9430036H15Rik, Carp1 |
Location: |
Chr.10:62743928-62792368 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccar1 | NM_026201.3 | 25 | 3775 | NP_080477.1 |
| AF465615.1 | 25 | 3666 | AAO17318.1 | |
| AK077151.1 | 9 | 1476 | BAC36646.1 | |
| AK078424.1 | 11 | 1752 | BAC37267.1 | |
| BC079652.1 | 25 | 3749 | AAH79652.1 |
More Information
Set Membership: |
|

Genomic Map