Assay Details
Target Gene Details
Entrez Gene ID: | 67629 |
Gene Name: | SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae) |
Gene Aliases: |
2410030K01Rik, AV109292, Spbc24 |
Location: |
Chr.9:21755442-21760452 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spc24 | NM_026282.5 | 5 | 639 | NP_080558.1 |
| AK011728.1 | 5 | 657 | BAB27804.1 | |
| AK031680.1 | 3 | 1553 | BAC27510.1 | |
| AK075950.1 | 5 | 640 | BAC36077.1 | |
| AK153185.1 | BAE31786.1 | |||
| AK167877.1 | BAE39891.1 | |||
| BC086683.1 | ||||
| BC116238.1 | ||||
| BC116239.1 |

Genomic Map