Assay Details
Target Gene Details
Entrez Gene ID: | 16798 |
Gene Name: | large tumor suppressor |
Gene Aliases: |
AW208599 |
Location: |
Chr.10:7681209-7716461 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lats1 | NM_010690.1 | 5 | 2577 | NP_034820.1 |
| AF104414.1 | 2 | 1702 | AAD16883.1 | |
| BC158092.1 | 4 | 2423 | ||
| BC158123.1 | 5 | 2572 |
More Information
Set Membership: |
|

Genomic Map