Assay Details
Target Gene Details
Entrez Gene ID: | 71137 |
Gene Name: | regulatory factor X, 4 (influences HLA class II expression) |
Gene Aliases: |
4933412G19Rik, NYD-sp10 |
Location: |
Chr.10:84756048-84906538 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx4 | NM_001024918.1 | 18 | 3417 | NP_001020089.1 |
| NM_027689.3 | 14 | 2749 | NP_081965.2 | |
| AB086957.1 | 18 | 3271 | BAD07408.1 | |
| AB089184.1 | 14 | 2750 | BAD07409.1 | |
| AK034131.1 | 18 | 3404 | BAC28598.1 | |
| AK132673.1 | 15 | 2812 | BAE21294.1 | |
| AY342003.1 | 18 | 3282 | AAQ17042.1 |
More Information
Set Membership: |
|

Genomic Map